missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ HACCP Flat Tray with Lid and Grid

Product Code. 16490821
Change view
Click to view available options
Color:
Blue
Green
White
Capacity (Metric):
3 L
8 L
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Capacity (Metric) Unit Size
16490821 Green 3 L Each
15999110 White 3 L Each
16480821 Blue 3 L Each
15909120 White 8 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16490821 Supplier Gilac™ Supplier No. G190136
  • ONLINE EXCLUSIVE PRICE
    £16.49
    £12.43 / Pack of 1
    Save £4.06
    24% Off
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HACCP Flat Tray can be used as a defrosting tray due to the wide dividing grid. Complete package with tray, shelf, and lid.

  • Flat Tray, Complete package: Tray + Shelf + Lid
  • Dimensions suitable for refrigerators
  • Can be used as:
    • defrosting trays thanks to its wide dividing grid that separates the ice water from the product being defrosted
    • preparation container for products that drip ingredients tray
    • containers for preserving comparison samples
    • fermenting and decanting containers
  • Nesting and stacking with lid

Specifications

Type Food Bin
Material HDPE
Color Green
Shape Rectangular
Capacity (English) 101.4 oz.
Capacity (Metric) 3 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.