missing translation for 'onlineSavingsMsg'
Learn More

H-ILE-PHE-OH, TRC

Product Code. 30389397
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30389397 50 mg
30398230 100 mg
30393867 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389397 Supplier Toronto Research Chemicals Supplier No. TRCI94208550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22951-98-0
Molecular Formula C15H22N2O3
Molecular Weight (g/mol) 278.35
Synonym Isoleucylphenylalanine, N-L-isoleucyl-L-Phenylalanine, L-Isoleucyl-L-phenylalanine, Ile-Phe
IUPAC Name (2S)-2-[[(2S,3S)-2-amino-3-methylpentanoyl]amino]-3-phenylpropanoic acid
SMILES CC[C@H](C)[C@H](N)C(=O)N[C@@H](Cc1ccccc1)C(=O)O

Specifications

InChI Formula InChI=1S/C15H22N2O3/c1-3-10(2)13(16)14(18)17-12(15(19)20)9-11-7-5-4-6-8-11/h4-8,10,12-13H,3,9,16H2,1-2H3,(H,17,18)(H,19,20)/t10-,12-,13-/m0/s1
Quantity 50 mg
Formula Weight 278.163
Shipping Condition Room Temperature
Chemical Name or Material H-ILE-PHE-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.