CAS RN 22951-98-0
CAS RN 22951-98-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
H-ILE-PHE-OH, TRC
CAS: 22951-98-0 Molecular Formula: C15H22N2O3 Molecular Weight (g/mol): 278.35 Synonym: Isoleucylphenylalanine,N-L-isoleucyl-L-Phenylalanine, L-Isoleucyl-L-phenylalanine, Ile-Phe IUPAC Name: (2S)-2-[[(2S,3S)-2-amino-3-methylpentanoyl]amino]-3-phenylpropanoic acid SMILES: CC[C@H](C)[C@H](N)C(=O)N[C@@H](Cc1ccccc1)C(=O)O