missing translation for 'onlineSavingsMsg'
Learn More

H-Glu(otbu)-nh2 hydrochloride, TRC

Product Code. 30240888
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240888 500 mg 500mg
30240594 250 mg 250mg
30240882 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240888 Supplier Toronto Research Chemicals Supplier No. TRCG598243500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108607-02-9
Molecular Formula C9H18N2O3 .HCl
Molecular Weight (g/mol) 238.71
Synonym (4S)-4,5-diamino-5-oxo-1,1-dimethylethyl Ester Pentanoic Acid Monohydrochloride;
IUPAC Name tert-butyl (4S)-4,5-diamino-5-oxopentanoate;hydrochloride
SMILES Cl.CC(C)(C)OC(=O)CC[C@H](N)C(=O)N

Specifications

InChI Formula InChI=1S/C9H18N2O3.ClH/c1-9(2,3)14-7(12)5-4-6(10)8(11)13;/h6H,4-5,10H2,1-3H3,(H2,11,13);1H/t6-;/m0./s1
Quantity 500 mg
Formula Weight 238.1084
Shipping Condition Room Temperature
Chemical Name or Material H-Glu(otbu)-nh2 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.