CAS RN 108607-02-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
H-Glu(otbu)-nh2 hydrochloride, TRC
CAS: 108607-02-9 Molecular Formula: C9H18N2O3 .HCl Molecular Weight (g/mol): 238.71 Synonym: (4S)-4,5-diamino-5-oxo-1,1-dimethylethyl Ester Pentanoic Acid Monohydrochloride; IUPAC Name: tert-butyl (4S)-4,5-diamino-5-oxopentanoate;hydrochloride SMILES: Cl.CC(C)(C)OC(=O)CC[C@H](N)C(=O)N