missing translation for 'onlineSavingsMsg'
Learn More

H-Asp-Leu-OH, TRC

Product Code. 30401921
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401921 10 mg
30405888 100 mg
30408738 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401921 Supplier Toronto Research Chemicals Supplier No. TRCI12012310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3062-14-4
Molecular Formula C10H18N2O5
Molecular Weight (g/mol) 246.26
Synonym L-alpha-Aspartyl-L-leucine, 141: PN: WO2011103105 SEQID: 152 unclaimed protein, 189: PN: EP2161028 PAGE: 10 claimed protein21: PN: WO2012158678 SEQID: 25 claimed sequence375: PN: WO2011130354 SEQID: 379 unclaimed protein53: PN: US20110305663 SEQID: 53 unclaimed protein703: PN: EP2071334 SEQID: 748 claimed protein703: PN: WO2009077864 SEQID: 748 claimed proteinL-Aspartyl-L-leucineNSC 332636alpha-Aspartylleucine
IUPAC Name (2S)-2-[[(2S)-2-amino-3-carboxypropanoyl]amino]-4-methylpentanoic acid
SMILES CC(C)C[C@H](NC(=O)[C@@H](N)CC(=O)O)C(=O)O

Specifications

InChI Formula InChI=1S/C10H18N2O5/c1-5(2)3-7(10(16)17)12-9(15)6(11)4-8(13)14/h5-7H,3-4,11H2,1-2H3,(H,12,15)(H,13,14)(H,16,17)/t6-,7-/m0/s1
Quantity 10 mg
Formula Weight 246.1216
Shipping Condition Room Temperature
Chemical Name or Material H-Asp-Leu-OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.