CAS RN 3062-14-4
CAS RN 3062-14-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
H-Asp-Leu-OH, TRC
CAS: 3062-14-4 Molecular Formula: C10H18N2O5 Molecular Weight (g/mol): 246.26 Synonym: L-alpha-Aspartyl-L-leucine,141: PN: WO2011103105 SEQID: 152 unclaimed protein,189: PN: EP2161028 PAGE: 10 claimed protein21: PN: WO2012158678 SEQID: 25 claimed sequence375: PN: WO2011130354 SEQID: 379 unclaimed protein53: PN: US20110305663 SEQID: 53 unclaimed protein703: PN: EP2071334 SEQID: 748 claimed protein703: PN: WO2009077864 SEQID: 748 claimed proteinL-Aspartyl-L-leucineNSC 332636alpha-Aspartylleucine IUPAC Name: (2S)-2-[[(2S)-2-amino-3-carboxypropanoyl]amino]-4-methylpentanoic acid SMILES: CC(C)C[C@H](NC(=O)[C@@H](N)CC(=O)O)C(=O)O