missing translation for 'onlineSavingsMsg'
Learn More

H-2D[d] Mouse anti-Mouse, FITC, Clone: 34-2-12, BD

Mouse Monoclonal Antibody

£88.17 - £375.78

Specifications

Antigen H-2D[d]
Clone 34-2-12
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15851549
View Documents
BD Biosciences
562001
50 μg
ONLINE EXCLUSIVE PRICE
£92.05
£88.17 / Each
Save £3.88
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15887288
View Documents
BD Biosciences
553579
0.5 mg
ONLINE EXCLUSIVE PRICE
£391.00
£375.78 / Each
Save £15.22
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The 34-2-12 antibody (also known as 34-2-12S) recognizes the α3 domain of the H-2D[d]. The binding of the antibody to its epitope is independent of the α1 and α2 domains and β2 microglobulin. It cross-reacts with cells of the C3H.LG/Ckc strain. Reactivity with other haplotypes (eg, b, f, k, p, q, r, s) has not been observed. Soluble mAb 34-2-12 blocks binding of the Ly-49A-expressing T lymphoma EL4 to immobilized H-2D[d]. However, further studies utilizing this mAb indicate that the α3 domain is not involved in the interaction between Ly-49A, or Ly-49G2, and H-2D[d].

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Specifications

Specifications

H-2D[d]
0.5mg/mL
Monoclonal
Mouse
Mouse
(C57BL/6 x DBA/2)F1 mouse splenocytes
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
34-2-12
Flow Cytometry
FITC
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
IgG2a κ
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.