missing translation for 'onlineSavingsMsg'
Learn More

H-2D[d] Mouse anti-Mouse, FITC, Clone: 34-2-12, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 553579

 View more versions of this product

Product Code. 15887288

  • ONLINE EXCLUSIVE PRICE
    £391.00
    £375.78 / Each
    Save £15.22
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 34-2-12 antibody (also known as 34-2-12S) recognizes the α3 domain of the H-2D[d]. The binding of the antibody to its epitope is independent of the α1 and α2 domains and β2 microglobulin. It cross-reacts with cells of the C3H.LG/Ckc strain. Reactivity with other haplotypes (eg, b, f, k, p, q, r, s) has not been observed. Soluble mAb 34-2-12 blocks binding of the Ly-49A-expressing T lymphoma EL4 to immobilized H-2D[d]. However, further studies utilizing this mAb indicate that the α3 domain is not involved in the interaction between Ly-49A, or Ly-49G2, and H-2D[d]. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 34-2-12
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: (C57BL/6 x DBA/2)F1 mouse splenocytes

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

H-2D[d]
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
(C57BL/6 x DBA/2)F1 mouse splenocytes
0.5 mg
Immunology
Mouse
IgG2a κ
Flow Cytometry
34-2-12
FITC
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.