missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GYPC Recombinant Rabbit Monoclonal Antibody (23GB6390), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | GYPC |
|---|---|
| Clone | 23GB6390 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089023
![]() |
Invitrogen™
MA553034 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087719
|
Invitrogen™
MA553035 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GYPC Recombinant Monoclonal antibody specifically detects GYPC in Human samples. It is validated for Western BlotSpecifications
| GYPC | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P04921 | |
| 2995 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6390 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 0610037F22Rik; CD236; CD236R; GE; Gerbich antigen; GLPC; Glycoconnectin; glycophorin C; glycophorin C (Gerbich blood group); glycophorin-C; Glycophorin-D; glycoprotein beta; GPC; GPD; GPYD; GYPC; GYPD; MGC117309; MGC126191; MGC126192; PAS-2; PAS-2'; Sialoglycoprotein D | |
| A synthesized peptide derived from human Glycophorin C (80-128AA). | |
| Primary | |
| -20°C | |
| GYPC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title