missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GYPC Recombinant Rabbit Monoclonal Antibody (23GB6390), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553035
This item is not returnable.
View return policy
Description
Glycophorin C (GYPC) is an integral membrane glycoprotein. It is a minor species carried by human erythrocytes, but plays an important role in regulating the mechanical stability of red cells. A number of glycophorin C mutations have been described. The Gerbich and Yus phenotypes are due to deletion of exon 3 and 2, respectively. The Webb and Duch antigens, also known as glycophorin D, result from single point mutations of the glycophorin C gene. The glycophorin C protein has very little homology with glycophorins A and B.
Specifications
| GYPC | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GYPC | |
| 0610037F22Rik; CD236; CD236R; GE; Gerbich antigen; GLPC; Glycoconnectin; glycophorin C; glycophorin C (Gerbich blood group); glycophorin-C; Glycophorin-D; glycoprotein beta; GPC; GPD; GPYD; GYPC; GYPD; MGC117309; MGC126191; MGC126192; PAS-2; PAS-2'; Sialoglycoprotein D | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 2995 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 23GB6390 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P04921 | |
| GYPC | |
| A synthesized peptide derived from human Glycophorin C (80-128AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction