missing translation for 'onlineSavingsMsg'
Learn More

Robu™ Guko Rubber Gasket

Robu™ Guko Rubber Gasket is a conical gasket suitable for use with filter funnels.

Brand:  Robu™ 21856

Additional Details : Weight : 0.17580kg

 View more versions of this product

Product Code. 16883036

  • £22.10 / Pack of 5
Estimated Shipment: 16-05-2024
to see stock.
Add to basket

Description

Description

  • Guko
  • Single
  • Conical gasket
  • Suitable for filter funnels
Specifications

Specifications

Single Gasket
Rubber
Gasket
48 mm outer, 32 mm inner
Funnel
Conical
35 mm
64 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers