missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanylate kinase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £301.00
Specifications
| Antigen | Guanylate kinase |
|---|---|
| Dilution | Western Blot 1:250-1: 1000, Flow (Intracellular) 1:500-1:1000 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18147381
|
Novus Biologicals
NBP2-23758SS |
0.025 mL |
£156.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18128351
|
Novus Biologicals
NBP2-23758 |
0.1 mL |
£301.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Guanylate kinase Polyclonal specifically detects Guanylate kinase in Human samples. It is validated for Western Blot, Flow (Intracellular).Specifications
| Guanylate kinase | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| EC 2.7.4.8, FLJ42686, FLJ43710, GMK, GMP kinase, guanylate kinase, guanylate kinase 1 | |
| GUK1 | |
| IgG | |
| Unpurified | |
| The amino acid sequence used as immunogen is 100% homologous in human, 94% homologous in chimpanzee and 66% homologous in monkey. This antibody will detect any Caspase-12 cleavage subunit containing amino acids 100-117. |
| Western Blot 1:250-1: 1000, Flow (Intracellular) 1:500-1:1000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide | |
| 2987 | |
| Within the range of amino acids 128-166 of human GUK1 protein were used as the immunogen. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title