missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guanylate kinase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-23758SS
This item is not returnable.
View return policy
Description
Guanylate kinase Polyclonal specifically detects Guanylate kinase in Human samples. It is validated for Western Blot, Flow (Intracellular).
Specifications
| Guanylate kinase | |
| Polyclonal | |
| Western Blot 1:250-1: 1000, Flow (Intracellular) 1:500-1:1000 | |
| EC 2.7.4.8, FLJ42686, FLJ43710, GMK, GMP kinase, guanylate kinase, guanylate kinase 1 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| 2987 | |
| Human | |
| Antisera |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| 0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide | |
| GUK1 | |
| Within the range of amino acids 128-166 of human GUK1 protein were used as the immunogen. | |
| 0.025 mL | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, 94% homologous in chimpanzee and 66% homologous in monkey. This antibody will detect any Caspase-12 cleavage subunit containing amino acids 100-117. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction