missing translation for 'onlineSavingsMsg'
Learn More

Guanosine-5'-triphosphate Trisodium Salt (Technical Grade), TRC

Product Code. 30240759
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240759 50 mg 50mg
30240626 25 mg 25mg
30240475 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240759 Supplier Toronto Research Chemicals Supplier No. TRCG83791050MG
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36051-31-7
Molecular Formula C10H13N5Na3O14P3
Molecular Weight (g/mol) 588.94
Synonym Guanosine 5'-(tetrahydrogen Triphosphate), Trisodium Salt, 5'-GTP Di-Na Salt, Trisodium 5'-GTP, GTP Trisodium Salt
IUPAC Name trisodium;[[[(2R,3S,4R,5R)-5-(2-amino-6-oxo-1H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl]oxy-oxidophosphoryl] hydrogen phosphate
SMILES [Na+].[Na+].[Na+].NC1=Nc2c(ncn2[C@@H]3O[C@H](COP(=O)([O-])OP(=O)([O-])OP(=O)(O)[O-])[C@@H](O)[C@H]3O)C(=O)N1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H16N5O14P3.3Na/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(27-9)1-26-31(22,23)29-32(24,25)28-30(19,20)21;;;/h2-3,5-6,9,16-17H,1H2,(H,22,23)(H,24,25)(H2,19,20,21)(H3,11,13,14,18);;;/q;3*+1/p-3/t3-,5-,6-,9-;;;/m1.../s1
Formula Weight 588.9365
Chemical Name or Material Guanosine-5'-triphosphate Trisodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.