missing translation for 'onlineSavingsMsg'
Learn More

Guanosine-5'-Triphosphate Trisodium Salt 98-100%, MP Biomedicals™

Product Code. p-4640976 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 11423751

Brand: MP Biomedicals 0215121680

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 36051-31-7
Molecular Formula C10H16MgN5O14P3
Molecular Weight (g/mol) 547.49
MDL Number MFCD00077781,MFCD00077781
InChI Key NMQPZZLDIUETGE-GWTDSMLYSA-N
Synonym 5'-gtp trisodium salt, 5 acute-gtp trisodium salt
PubChem CID 92044363
SMILES [Mg].NC1=NC(=O)C2=C(N1)N(C=N2)[C@@H]1O[C@H](COP(O)(=O)OP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O

Specifications

Color White
Assay Percent Range 98 to 100%
Quantity 100 mg
IUPAC Name trisodium;[[(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate
Formula Weight 588.2 (Anhydrous)
Physical Form Powder
Chemical Name or Material Guanosine-5'-Triphosphate Trisodium Salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.