missing translation for 'onlineSavingsMsg'
Learn More

Guanosine-5'-Triphosphate Trisodium Salt 98-100%, MP Biomedicals™

Product Code. 11423751 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423751 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11423751 Supplier MP Biomedicals Supplier No. 0215121680
  • ONLINE EXCLUSIVE PRICE
    £203.00
    £190.05 / 100mg
    Save £12.95
    6% Off
 Restricted Item
Estimated Shipment: 15-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 36051-31-7
Molecular Formula C10H16MgN5O14P3
Molecular Weight (g/mol) 547.49
MDL Number MFCD00077781,MFCD00077781
InChI Key NMQPZZLDIUETGE-GWTDSMLYSA-N
Synonym 5'-gtp trisodium salt, 5 acute-gtp trisodium salt
PubChem CID 92044363
SMILES [Mg].NC1=NC(=O)C2=C(N1)N(C=N2)[C@@H]1O[C@H](COP(O)(=O)OP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O

Specifications

Color White
Assay Percent Range 98 to 100%
Quantity 100 mg
IUPAC Name trisodium;[[(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate
Formula Weight 588.2 (Anhydrous)
Physical Form Powder
Chemical Name or Material Guanosine-5'-Triphosphate Trisodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.