missing translation for 'onlineSavingsMsg'
Learn More

Guanosine-5'-triphosphate Disodium Salt, ≥95%, MP Biomedicals™

Product Code. p-4405806 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 11441132

Brand: MP Biomedicals 0219521391

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Guanosine-5'-triphosphate Disodium Salt, ≥95% is a substrate for RNA polymerases.

  • Guanosine 5′-triphosphate (GTP), a purine trinucleotide, is a substrate for RNA polymerases and a wide range of GTPase including G-protein coupled receptor (GPCR) GTPases and cell signaling and cycling associated guanine nucleotide exchange factors (GEF).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36051-31-7
Molecular Formula C10H16MgN5O14P3
Molecular Weight (g/mol) 547.49
MDL Number MFCD03410297
InChI Key NMQPZZLDIUETGE-GWTDSMLYSA-N
Synonym guanosine 5'-triphosphate, guanosine-5'-triphosphate disodium salt dihydrate gtp, guanosine-triphosphate, guanosine 5'-triphosphate 4-, gtp 4-, 2r,3s,4r,5r-5-2-amino-6-hydroxy-9h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methyl phosphonato oxy phosphonatooxy phosphinate, 2r,3s,4r,5r-5-2-amino-6-oxo-3h-purin-9-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl phosphate
PubChem CID 131676145
SMILES [Mg].NC1=NC(=O)C2=C(N1)N(C=N2)[C@@H]1O[C@H](COP(O)(=O)OP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]1O

Specifications

Absorbance λ max (water): 252 ± 3 nm
Color White
Assay Percent Range ≥95%
Quantity 1 g
IUPAC Name disodium;(2R,3R,4S,5R)-2-(2-amino-6-oxo-3H-purin-9-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate
Formula Weight 567.2
Percent Purity ≥95%
Physical Form Powder
Chemical Name or Material Guanosine-5'-triphosphate disodium salt, For HPLC analysis
compliance-icons
Product Identifier
  • Guanosine-5'-triphosphate Disodium Salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : guanosine 5'-(disodium dihydrogentriphosphate)

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.