missing translation for 'onlineSavingsMsg'
Learn More

Melford Guanidine Thiocyanate

Product Code. 30018311
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018311 500 g 500g
30018310 1 kg 1kg
30018309 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018311 Supplier Melford Supplier No. G54000500.0
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protein denaturant used for purifying proteins and nucleic acids.

Protein denaturant used for purifying proteins and nucleic acids. Toxicity: LD50

Specifications

CAS 593-84-0
Molecular Formula CH5N3-HSCN
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.