missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTPBP9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£213.00 - £459.00
Specifications
| Antigen | GTPBP9 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18491781
|
Novus Biologicals
NBP1-89725-25ul |
25 μL |
£213.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18412091
|
Novus Biologicals
NBP1-89725 |
0.1 mL |
£459.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GTPBP9 Polyclonal antibody specifically detects GTPBP9 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDownSpecifications
| GTPBP9 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Rabbit | |
| Proteases & Other Enzymes | |
| PBS (pH 7.2) and 40% Glycerol | |
| 29789 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockdown Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp313H1942, DNA damage-regulated overexpressed in cancer 45 protein, DOC45, EC 3.6.3, EC 3.6.3.-, GBP45, GTP-binding protein 9, GTP-binding protein 9 (putative), GTP-binding protein PTD004, GTPBP9, homologous yeast-44.2 protein, Obg-like ATPase 1, PTD004 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LTNSQASAENFPFCTIDPNESRVPVPDERFDFLCQYHKPASKIPAFLNVVDIAGLVKGAHNGQGLGNAFLSHISACDGIFHLTRAFED | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title