missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTPBP9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57453
This item is not returnable.
View return policy
Description
GTPBP9 Polyclonal specifically detects GTPBP9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GTPBP9 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DKFZp313H1942, DNA damage-regulated overexpressed in cancer 45 protein, DOC45, EC 3.6.3, EC 3.6.3.-, GBP45, GTP-binding protein 9, GTP-binding protein 9 (putative), GTP-binding protein PTD004, GTPBP9, homologous yeast-44.2 protein, Obg-like ATPase 1, PTD004 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Rat: 100%; Human: 100%; Xenopus: 100%; Western clawed frog: 100%; Chicken: 100%; Sumatran orangutan: 100%; Green puffer: 100%; Bovine: 100%; Mouse: 100%; Atlantic salmon: 92%; Rainbow smelt: 92%; Zebrafish: 92%; Fruit fly: 85%; Argulus sp. Arg2: 85%; Southern house mosquito: 78%; Pea aphid: 78%; Hanseniella sp. 'Han2': 78%; Orchesella imitari: 78%; Tomocerus sp. 'Tom2': 78%; Savannah tsetse fly: 78%; African malaria mosquito: 78%; Giant whipscorpion: 78%;. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q9NTK5 | |
| OLA1 | |
| Synthetic peptides corresponding to GTPBP9 (GTP-binding protein 9 (putative)) The peptide sequence was selected from the N terminal of GTPBP9. Peptide sequence MIGPIIDKLEKVAVRGGDKKLKPEYDIMCKVKSWVIDQKKPVRFYHDWND. | |
| 100 μL | |
| Proteases & Other Enzymes | |
| 29789 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction