missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTPBP4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13897-1-AP-150UL
This item is not returnable.
View return policy
Description
GTP-binding proteins are GTPases and function as molecular switches that can flip between two states: active, when GTP is bound, and inactive, when GDP is bound. ′Active′ in this context usually means that the molecule acts as a signal to trigger other events in the cell. When an extracellular ligand binds to a G-protein-linked receptor, the receptor changes its conformation and switches on the trimeric G proteins that associate with it by causing them to eject their GDP and replace it with GTP. The switch is turned off when the G protein hydrolyzes its own bound GTP, converting it back to GDP. But before that occurs, the active protein has an opportunity to diffuse away from the receptor and deliver its message for a prolonged period to its downstream target.Specifications
| GTPBP4 | |
| Polyclonal | |
| Unconjugated | |
| GTPBP4 | |
| CRFG, GTP binding protein 4, GTP binding protein NGB, GTPBP4, NGB, NOG1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23560, 69237 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99ME9, Q9BZE4 | |
| Gtpbp4 | |
| GTPBP4 Fusion Protein Ag4859 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction