missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTPBP10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-52959
This item is not returnable.
View return policy
Description
GTPBP10 Polyclonal specifically detects GTPBP10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GTPBP10 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DKFZP686A10121, FLJ38242, GTP-binding protein 10, GTP-binding protein 10 (putative), MGC104191, ObgH2, Protein obg homolog 2 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 85865 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| A4D1E9 | |
| GTPBP10 | |
| Synthetic peptides corresponding to GTPBP10(GTP-binding protein 10 (putative)) The peptide sequence was selected from the middle region of GTPBP10. Peptide sequence IILLTKELELYKEELQTKPALLAVNKMDLPDAQDKFHELMSQLQNPKDFL. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Canine: 92%; Guinea pig: 92%; Chicken: 85%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction