missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF2I Recombinant Rabbit Monoclonal Antibody (23GB5950), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | GTF2I |
|---|---|
| Clone | 23GB5950 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089742
![]() |
Invitrogen™
MA553934 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088102
|
Invitrogen™
MA553935 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GTF2I Recombinant Monoclonal antibody specifically detects GTF2I in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| GTF2I | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P78347, Q5U2Y1, Q9ESZ8 | |
| 14886, 2969, 353256 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5950 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 6030441I21Rik; BAP135; BAP-135; Bruton tyrosine kinase-associated protein 135; BTKAP1; BTK-associated protein 135; BTK-associated protein, 135kD; DIWS; Diws1t; general transcription factor II I; general transcription factor IIi; general transcription factor II-I; Gtf2i; Gtf2ird1; GTFII-I; IB291; OTTHUMP00000209509; Spin; SRF-Phox1-interacting protein; tfiii; TFII-I; WBS; WBSCR6; Williams-Beuren syndrome chromosomal region 6 protein; Williams-Beuren syndrome chromosome region 6 | |
| A synthesized peptide derived from GTF2I (50-150AA). | |
| Primary | |
| -20°C | |
| GTF2I |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title