missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GTF2I Recombinant Rabbit Monoclonal Antibody (23GB5950), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553935
This item is not returnable.
View return policy
Description
This gene encodes a multifunctional phosphoprotein with roles in transcription and signal transduction. It is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at chromosome 7q11. 23. Alternative splicing results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 7, 13 and 21.
Specifications
| GTF2I | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GTF2I | |
| 6030441I21Rik; BAP135; BAP-135; Bruton tyrosine kinase-associated protein 135; BTKAP1; BTK-associated protein 135; BTK-associated protein, 135kD; DIWS; Diws1t; general transcription factor II I; general transcription factor IIi; general transcription factor II-I; Gtf2i; Gtf2ird1; GTFII-I; IB291; OTTHUMP00000209509; Spin; SRF-Phox1-interacting protein; tfiii; TFII-I; WBS; WBSCR6; Williams-Beuren syndrome chromosomal region 6 protein; Williams-Beuren syndrome chromosome region 6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14886, 2969, 353256 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5950 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P78347, Q5U2Y1, Q9ESZ8 | |
| GTF2I | |
| A synthesized peptide derived from GTF2I (50-150AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction