missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gtf2h5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121936
This item is not returnable.
View return policy
Description
Component of the general transcription and DNA repair factor IIH (TFIIH) core complex, which is involved in general and transcription-coupled nucleotide excision repair (NER) of damaged DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II. In NER, TFIIH acts by opening DNA around the lesion to allow the excision of the damaged oligonucleotide and its replacement by a new DNA fragment. In transcription, TFIIH has an essential role in transcription initiation. When the pre-initiation complex (PIC) has been established, TFIIH is required for promoter opening and promoter escape. Phosphorylation of the C-terminal tail (CTD) of the largest subunit of RNA polymerase II by the kinase module CAK controls the initiation of transcription. Necessary for the stability of the TFIIH complex and for the presence of normal levels of TFIIH in the cell.
Specifications
| Gtf2h5 | |
| Polyclonal | |
| Unconjugated | |
| GTF2H5 | |
| 2700017P07Rik; 2810432H05Rik; bA120J8.2; C6orf175; D17Wsu155e; General transcription factor IIH polypeptide 5; General transcription factor IIH subunit 5; general transcription factor IIH, polypeptide 5; GTF2H5; RGD1560991; si:dkey-12h9.4; TFB5; TFB5 ortholog; TFIIH; TFIIH basal transcription factor complex TTDA subunit; TFIIH basal transcription factor complex TTD-A subunit; TGF2H5; TTD; TTD3; TTDA; TTD-A; Unknown (protein for MGC:134263); Unknown (protein for MGC:153434); zgc:153434 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 404672 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q6ZYL4 | |
| GTF2H5 | |
| Met1-Lys71 (Accession Q6ZYL4), with N-terminal His Tag. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction