missing translation for 'onlineSavingsMsg'
Learn More

Cytiva GSTrap™ HP Columns

Product Code. 11713219 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 5 mL
5 x 1 mL
5 x 5 mL
Unit Size:
Each
Pack of 5
Product Code. Quantity unitSize
11713219 1 x 5 mL Each
10647455 5 x 1 mL Pack of 5
11565035 5 x 5 mL Pack of 5
3 options
This item is not returnable. View return policy

Product Code. 11713219

Brand: Cytiva 17528201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use to provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins. Cytiva Life Sciences™ GSTrap™ HP Columns are prepacked with Glutathione Sepharose™ High Performance medium.

  • One-step purification of GST-tagged proteins and other glutathione S-transferases or glutathione-dependent proteins
  • Glutathione coupled to Sepharose™ High Performance media
  • Column material: polypropylene
  • May be used with syringe, peristaltic pump or chromatography system
  • Purification process preserves antigenicity and function
  • Available as 1mL and 5mL columns
Media
  • Average particle size: 34μm
  • Ligand: Glutathione
  • Matrix: Highly cross-linked 6% agarose
  • Binding capacity: >7mg/mL
  • pH stability working range: 3 to 12

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.
5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 5 mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16 mm
Description GSTrap HP columns are prepacked with Glutathione Sepharose High Performance resin and provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins.
Flow Rate <20 mL/min.
Material Polypropylene
pH Range 3 to 12
Support Type 4% Highly Cross-linked Agarose
Content And Storage 4°C to 30°C, 20% Ethanol
Max. Operating Pressure 5 bar (0.5 MPa, 70 psi)
Particle Size 34 μm
Quantity 1 x 5 mL
For Use With (Application) Glutathione S-transferase (GST) tagged proteins produced using the pGEX series of expression vectors, Other glutathione S-transferases and glutathione-dependent proteins
Type Pre-packed Column
Ligand Type Glutathione
Show More Show Less
compliance-icons
Product Identifier
  • GSTrap HP 5 ml, 1 x 5 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.