missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTM3 Mouse, anti-Human, CoraLite Plus 488, Clone: 1E2B8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | GSTM3 |
|---|---|
| Clone | 1E2B8 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18056905
|
Proteintech
CL488-67634-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18095155
|
Proteintech
CL488-67634-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The GSTM3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GSTM3. This antibody recognizes human, pig antigen. The GSTM3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| GSTM3 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 2947 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1E2B8 | |
| Immunofluorescence | |
| Liquid | |
| Human, Pig | |
| Glutathione S transferase Mu 3, GST class mu 3, GST5, GSTB, GSTM3, GSTM3 3, GTM3, hGSTM3 3 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title