Learn More
GSTM3 Mouse, anti-Human, CoraLite Plus 488, Clone: 1E2B8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67634-100UL
Description
The GSTM3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GSTM3. This antibody recognizes human, pig antigen. The GSTM3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| GSTM3 | |
| 1E2B8 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Pig | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| Glutathione S transferase Mu 3, GST class mu 3, GST5, GSTB, GSTM3, GSTM3 3, GTM3, hGSTM3 3 | |
| 100 μL | |
| 2947 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |