Learn More
Cytiva GST MultiTrap™ 4B Plates
96-well filter plates for high-throughput screening and parallel purification of GST-tagged proteins
Brand:  Cytiva 28-4055-00
Code : GE
Additional Details : Weight : 0.52800kg
Product Code. 11753319
  • £832.00 / Pack of 4
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
Description
Description
  • Highly reproducible well-to-well and plate-to-plate results using manual or robotic systems
  • High purity in one step with high chemical stability
  • Allows loading of unclarified cell lysate samples directly into wells
  • Prepacked with Glutathione Sepharose media
Specifications
Specifications
GST MultiTrap FF and GST MultiTrap 4B are prepacked, 96-well filter plates for reproducible, high-throughput screening and rapid parallel purification of glutathione S-transferase (GST) tagged proteins.
4°C to 30°C, 20% Ethanol
Filter Plate
Glutathione Sepharose 4B
4 x 96-well Filter Plates