missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 9027
Description
Glucocorticoid receptor agonist (pIC50 = 8). Inhibits production of the proinflammatory mediator IL-6 in vivo.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 525.52 |
| Purity | >98% |
| Molecular Weight (g/mol) | 525.522 |
| Chemical Name or Material | GSK 9027 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?