CAS RN 1229096-88-1
CAS RN 1229096-88-1
IUPAC Name:
N-{4-[1-(4-fluorophenyl)-1H-indazol-5-yl]-3-(trifluoromethyl)phenyl}-1-phenylmethanesulfonamide
Synonyms:
N-{4-[1-(4-fluorophenyl)indazol-5-yl]-3-(trifluoromethyl)phenyl}-1-phenylmethanesulfonamide
Molecular Weight (g/mol):
525.52
Molecular Formula:
C27H19F4N3O2S
InChi Key:
DXBJGDVBQPEMOB-UHFFFAOYSA-N
SMILES:
FC1=CC=C(C=C1)N1N=CC2=CC(=CC=C12)C1=C(C=C(NS(=O)(=O)CC2=CC=CC=C2)C=C1)C(F)(F)F