missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK 9027

Product Code. 18046042
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18046042 10 mg 10mg
18073872 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18046042 Supplier Tocris Bioscience™ Supplier No. 4116/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glucocorticoid receptor agonist

Glucocorticoid receptor agonist (pIC50 = 8). Inhibits production of the proinflammatory mediator IL-6 in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1229096-88-1
Molecular Formula C27H19F4N3O2S

Specifications

Quantity 10 mg
Formula Weight 525.52
Purity >98%
Molecular Weight (g/mol) 525.522
Chemical Name or Material GSK 9027
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.