missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSC Recombinant Rabbit Monoclonal Antibody (23GB6470), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | GSC |
|---|---|
| Clone | 23GB6470 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088442
|
Invitrogen™
MA553962 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089348
|
Invitrogen™
MA553963 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GSC Recombinant Monoclonal antibody specifically detects GSC in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| GSC | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P56915, Q02591 | |
| 145258, 14836, 317715 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6470 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| GCS; goosecoid; goosecoid homeobox; goosecoid protein; GSC; Homeobox protein goosecoid; SAMS | |
| A synthesized peptide derived from human GSC (1-37AA). | |
| Primary | |
| -20°C | |
| GSC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title