missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSC Recombinant Rabbit Monoclonal Antibody (23GB6470), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553962
This item is not returnable.
View return policy
Description
This gene encodes a member of the bicoid subfamily of the paired (PRD) homeobox family of proteins. The encoded protein acts as a transcription factor and may be autoregulatory. A similar protein in mice plays a role in craniofacial and rib cage development during embryogenesis.
Specifications
| GSC | |
| Recombinant Monoclonal | |
| 0.575 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P56915, Q02591 | |
| GSC | |
| A synthesized peptide derived from human GSC (1-37AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6470 | |
| Unconjugated | |
| GSC | |
| GCS; goosecoid; goosecoid homeobox; goosecoid protein; GSC; Homeobox protein goosecoid; SAMS | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 145258, 14836, 317715 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction