missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Growth Cone Antibody (2G13), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
£160.00 - £386.00
Specifications
| Antigen | Growth Cone |
|---|---|
| Clone | 2G13 |
| Dilution | Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18289131
|
Novus Biologicals
NB600-785-0.25ML |
0.25 mL |
£160.00
0.25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18719613
|
Novus Biologicals
NB600-785 |
1 mL |
£386.00
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Growth Cone Monoclonal specifically detects Growth Cone in Mouse, Rat, Chicken samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Growth Cone | |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Rat, Mouse, Chicken | |
| Embryonic chick tectal membranes | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 2G13 | |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Neuroscience | |
| Tissue Culture Supernatant with 0.09% Sodium Azide | |
| IgM | |
| Tissue culture supernatant | |
| This is useful as a general reagent for the detection of axonal growth cones in tissue sections and potentially a key marker in neuronal development research. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title