missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Growth Cone Antibody (2G13), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB600-785
This item is not returnable.
View return policy
Description
Growth Cone Monoclonal specifically detects Growth Cone in Mouse, Rat, Chicken samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Growth Cone | |
| Monoclonal | |
| Unconjugated | |
| Tissue Culture Supernatant with 0.09% Sodium Azide | |
| Embryonic chick tectal membranes | |
| 1 mL | |
| Neuroscience | |
| This is useful as a general reagent for the detection of axonal growth cones in tissue sections and potentially a key marker in neuronal development research. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgM |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 2G13 | |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Mouse | |
| Tissue culture supernatant | |
| RUO | |
| Primary | |
| Mouse, Rat, Chicken | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction