missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Group B Strep Selective Enrichment Broth
Description
Group B Strep Selective Enrichment Broth is used to selectively enrich and recover Group B Streptococcus (Streptococcus agalactiae) from clinical specimens, such as vaginal or rectal swabs.
Specifications
Specifications
| Color | Straw |
| Content And Storage | 2°C to 25°C |
| Format | Plastic universal bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Microbiology Media |
| Fill Weight | 10.0 to 10.3 g |
| Packaging Type | 24 Plastic Universal Bottles with screw cap closures in a box |
| Quantity | 24/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?