missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRK3 Recombinant Rabbit Monoclonal Antibody (23GB6300), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | GRK3 |
|---|---|
| Clone | 23GB6300 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087793
![]() |
Invitrogen™
MA553014 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087803
|
Invitrogen™
MA553015 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRK3 Recombinant Monoclonal antibody specifically detects GRK3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| GRK3 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P26819, P35626, Q3UYH7 | |
| 157, 25372, 320129 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6300 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 4833444A01Rik; ADRBK2; Adrbk-2; adrenergic receptor kinase, beta 2; adrenergic receptor kinase, beta 2 (G-protein-linked receptor kinase); adrenergic, beta, receptor kinase 2; AI851927; AW551196; BARK2; Bark-2; beta ARK2; beta-adrenergic receptor kinase 2; Beta-ARK-2; G protein-coupled receptor kinase 3; G-protein-coupled receptor kinase 3; Grk3 | |
| A synthesized peptide derived from human GRK3 (620-688AA). | |
| Primary | |
| -20°C | |
| GRK3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title