missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRK3 Recombinant Rabbit Monoclonal Antibody (23GB6300), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553015
This item is not returnable.
View return policy
Description
ADRBK2, or adrenergic beta receptor kinase 2 is a serine/threonine kinase that is thought to act as a regulator of receptor function. Overall, the ADRBK2 enzyme, also known as GRK3, has 85% amino acid similarity with ADRBK1, with the protein kinase catalytic domain having 95% similarity. The ADRBK2 mRNA is approximately 8 kilobases with a distribution similar to that of ADRBK1. These data suggest the existence of a family of receptor kinases which may serve broadly to regulate receptor function.
Specifications
| GRK3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| GRK3 | |
| 4833444A01Rik; ADRBK2; Adrbk-2; adrenergic receptor kinase, beta 2; adrenergic receptor kinase, beta 2 (G-protein-linked receptor kinase); adrenergic, beta, receptor kinase 2; AI851927; AW551196; BARK2; Bark-2; beta ARK2; beta-adrenergic receptor kinase 2; Beta-ARK-2; G protein-coupled receptor kinase 3; G-protein-coupled receptor kinase 3; Grk3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 157, 25372, 320129 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6300 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P26819, P35626, Q3UYH7 | |
| GRK3 | |
| A synthesized peptide derived from human GRK3 (620-688AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction