missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK2 Antibody (3F8), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£173.00 - £435.00
Specifications
| Antigen | GRK2 |
|---|---|
| Clone | 3F8 |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18610366
|
Novus Biologicals
NBP2-37611-0.025ml |
0.025 mL |
£173.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18728173
|
Novus Biologicals
NBP2-37611 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GRK2 Monoclonal specifically detects GRK2 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| GRK2 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Rat, Primate, Monkey | |
| adrenergic, beta, receptor kinase 1, BARK, BARK1beta-ARK-1, Beta-ARK-1, BETA-ARK1, EC 2.7.11, EC 2.7.11.15, FLJ16718, G-protein coupled receptor kinase 2, GRK2beta-adrenergic receptor kinase 1 | |
| GRK2 | |
| IgG1 | |
| Unpurified |
| 3F8 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, Mitochondrial Markers, Neuroscience, Protein Kinase, Signal Transduction | |
| Ascites with 0.03% Sodium Azide | |
| 156 | |
| Purified recombinant fragment of human GRK2 expressed in E. Coli. Sequence is within AA 389-583 | |
| Primary | |
| 80 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title