missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK2 Antibody (3F8), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-37611-0.025ml
This item is not returnable.
View return policy
Description
GRK2 Monoclonal specifically detects GRK2 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| GRK2 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| GRK2 | |
| Purified recombinant fragment of human GRK2 expressed in E. Coli. Sequence is within AA 389-583 | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Primate, Monkey | |
| Ascites |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 3F8 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| adrenergic, beta, receptor kinase 1, BARK, BARK1beta-ARK-1, Beta-ARK-1, BETA-ARK1, EC 2.7.11, EC 2.7.11.15, FLJ16718, G-protein coupled receptor kinase 2, GRK2beta-adrenergic receptor kinase 1 | |
| Mouse | |
| 80 kDa | |
| 0.025 mL | |
| Cancer, Cellular Markers, Mitochondrial Markers, Neuroscience, Protein Kinase, Signal Transduction | |
| 156 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction