Learn More
Fritsch™ Grinding Sets
Brand: Fritsch™ 46.2080.00
Code : 04
Additional Details : Weight : 6.10000kg
Specifications
Grinding Set | |
Mortar Grinder Pulverisette 2 | |
Hardmetal Tungsten Carbide |
Hard, medium-hard, and abrasive samples | |
Highly centralized mortar bowl rimmed in a plastic frame, pestle, and scraper |