Learn More
SPEX Certiprep™ Grinding cylinder Geno/Grinder Ceramic
Shop All SPEX Certiprep Products
Click to view available options
Dimensions (L x W):
3/8 x 7/8 in.
5/32 x 5/16 in.
Unit Size:
Pack of 100
Description
- Chemically inert
- Angle-cut end help shear the sample matrix and ensure good homogenisation and maximise extraction rates
Specifications
Specifications
| Capacity (Metric) | 5 or 15 mL |
| Dimensions (L x W) | 5/32 x 5/16 in. |
| For Use With (Application) | Two cylinders are used per sample vial |
| Packaging Quantity | 100/bag |