missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRG (Groucho homolog) Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | GRG (Groucho homolog) |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
GRG (Groucho homolog) Polyclonal specifically detects GRG (Groucho homolog) in Mouse samples. It is validated for Western Blot.Specifications
| GRG (Groucho homolog) | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Signal Transduction, Wnt Signaling Pathway | |
| PBS buffer, 2% sucrose | |
| 166 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| AES-1, AES-2, Amino enhancer of split, amino-terminal enhancer of split, ESP1, Gp130-associated protein GAM, GRG, GRG5, Protein ESP1, Protein GRG, TLE5 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Mouse GRG (Groucho homolog) (NP_034477). Peptide sequence QSRHSGSSHLPQQLKFTTSDSCDRIKDEFQLLQAQYHSLKLECDKLASEK | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title