missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRG (Groucho homolog) Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
GRG (Groucho homolog) Polyclonal specifically detects GRG (Groucho homolog) in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | GRG (Groucho homolog) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | AES-1, AES-2, Amino enhancer of split, amino-terminal enhancer of split, ESP1, Gp130-associated protein GAM, GRG, GRG5, Protein ESP1, Protein GRG, TLE5 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Mouse GRG (Groucho homolog) (NP_034477). Peptide sequence QSRHSGSSHLPQQLKFTTSDSCDRIKDEFQLLQAQYHSLKLECDKLASEK |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?