missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek Green Test Dye
Shop All Agilent BioTek Products
Click to view available options
Product Type:
10X Concentrate Test Dye
50x Concentrate Test Dye
Test Dye
Unit Size:
125mL
Specifications
Specifications
| Product Type | Test Dye |
| For Use With (Equipment) | Agilent BioTek Microplate Washers and Readers |
| For Use With (Application) | Test reagent injectors on all multi-mode readers during instrument qualification |
| Quantity | 125 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction