Learn More
MULTIROIR Gray Euro-Standard Solid Container
Product Code. 17296910
Click to view available options
Dimensions (L x W x H):
400 x 300 x 170 mm
400 x 300 x 230 mm
600 x 400 x 120 mm
600 x 400 x 170 mm
600 x 400 x 230 mm
600 x 400 x 290 mm
600 x 400 x 340 mm
600 x 400 x 450 mm
Unit Size:
Each
Product Code. 17296910
Brand: MULTIROIR SPK4017_GREY
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Euro standard plastic storage bin is compatible with all containers meeting European standards and in particular Euro pallets
  • The Euro standard plastic storage bin is compatible with all containers meeting European standards and in particular Euro pallets
  • The Euro standard container from Multiroir allows the storage, transport and supply of goods in manual or automated mode, indoors or outdoors
  • Manufactured in thick polypropylene and therefore undeformable and very robust
  • Standard silver gray color
  • Euro container with full walls
  • Resistant to acids and bases, cold and heat
  • Food grade
  • Ergonomic handles for optimal handling
  • Smooth walls for easy cleaning
Specifications
Type Storage Box
Material Polypropylene
Color Gray
Shape Rectangular
Includes Open handles
Dimensions (L x W x H) 400 x 300 x 170 mm