missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences Strep-Tactin™XT Superflow™ Column
Shop All IBA Lifesciences Products
Click to view available options
Capacity (Metric):
1 x 1 mL
1 x 5 mL
5 x 0.2 mL
1 x 10 mL
Quantity:
1 Column
5 Columns
Unit Size:
Each
Pack of 5
Description
- Strep-Tactin™XT Superflow™ columns contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™XT and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
- As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which leads to an outstanding purity of the target protein.
- In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™XT Superflow™ has a higher affinity for Strep-tag™ fusion proteins, which leads to an improved immobilization and purification.
- In addition, Strep-Tactin™XT Superflow™ allows the application of denaturing agents such as urea during the purification procedure.
- To reduce the waiting period during the sample and buffer application, the use of the application aid WET FRED is recommended.
Specifications
Specifications
| Capacity (Metric) | 1 x 5 mL |
| Quantity | 1 Column |
| For Use With (Application) | Protein Purification |
| Type | Gravity Flow Column |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction