missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences Gravity flow Strep-Tactin™ 4Flow™ high capacity column
Shop All IBA Lifesciences ProductsDescription
Specifications
Specifications
| Binding Capacity | 20 mg/mL resin |
| Capacity (Metric) | 5 mL |
| Description | 4% agarose, crosslinked |
| Particle Size | 50 to 150 μm |
| Form | Resin Slurry |
| Quantity | 5 mL |
| Protein Form | Tetrameric protein |
| Target | Proteins tagged with Twin-Strep-tag™; Strep-tag™II; cross reactivity: biotin |
| Type | Column |
| Final Product Type | Affinity chromatography equipment |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?